Principaux documents
1.52618
Supelco
Dust Cover
greener alternative
Replacement part
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.21
Niveau de qualité
Caractéristiques du produit alternatif plus écologique
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
Autre catégorie plus écologique
Température de stockage
2-30°C
Description générale
The Dust Cover is appropiate for a proper protection from dust designed for the TLC Explorer Documentation System (Cat. No. 1.52610.0001 ). The TLC Explorer Documentation System is designed to document and evaluate thin-layer chromatographic (TLC) plates. It offers a safe, fast and flexible way to visually inspect and digitally record images of TLC plates after their chromatographic development workflow in the laboratory. For detecting different substance classes, the device contains three different LED light sources (white light (VIS), UVA (366 nm) and UVC (254 nm), which illuminate the plates from above in direct-light mode. Recorded plate images can be evaluated to generate reports or export the data for further processing with other systems.
We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of greener alternatives. This product is a Design for Sustainability (DfS) developed product. Click here to view its DfS scorecard.