Principaux documents
Z743263
Julabo® bath tubing
tubing insulation, I.D. × L 18 mm × 1 m , for use with 12 mm I.D. reinforced tubing
About This Item
Code UNSPSC :
41103706
Nomenclature NACRES :
NB.77
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Julabo 8930412
D.I. × L
18 mm × 1 m , for use with 12 mm I.D. reinforced tubing
Description générale
Flexible connection of the application to the temperature control instrument for optimal use in the respective temperature range.
Informations légales
Julabo is a registered trademark of Julabo USA, Inc.