Principaux documents
Z742843
Kern PCD series precision balances, rechareable battery pack
About This Item
Code UNSPSC :
41111502
Nomenclature NACRES :
NB.71
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Kern PCD-A04
Description générale
Internal rechargeable NiMH battery pack PCD-A04 for se with Kern PCD precision balances. Operating time up to 24 h, charging time approx. 12 h, Auto-Off function preserves the rechargeable battery pack.