Principaux documents
Z742483
Prefilled tubes for BeadBlaster 24 Microtube and BeadBug homogenizer
2 ml tubes, with mixed format: 0.1 mm silica, 1.4 mm zirconium, and 4 mm silica
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.61
Caractéristiques
2 ml tubes
Conditionnement
pkg of 50
Fabricant/nom de marque
Benchmark Scientific D1034-MX
Description générale
Prefilled tubes; Compatible with all common bead homogenizers
Informations légales
BeadBlaster is a trademark of Benchmark Scientific, Inc.
Global Trade Item Number
RéférenceGTIN
Z742483-50EA4061835332984