Principaux documents
Z742415
High pressure adapter for Q-Tube
for 35 mL Q-Tube, max. pressure 180 psi
About This Item
Code UNSPSC :
51111800
Nomenclature NACRES :
NB.51
Matériaux
aluminum
Fabricant/nom de marque
Q Labtech 100-TA0025-350
Paramètres
-200 psi pressure (max)
180 psi max. pressure
Description générale
Pressure Adapter-35 mL-180 psi is designed to fit on 35 mL systems. This blue anodized aluminum adapter can hold up to 180 psi internal pressure. This higher threshold is ideal for hydrogenation and reactions involving ammonia hydroxide, methylamine, dimethylamine, formaldehyde, acetaldehyde, and hydrogen chloride.
Informations légales
Q-Tube is a trademark of Q Labtech LLC