Principaux documents
Z674672
Slide spinner for slide drying
AC/DC input 230 V AC, Schuko plug
Synonyme(s) :
slide dryer
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
MA.01
Fabricant/nom de marque
Labnet C1303-T-230V-EU
Paramètres
~4800 rpm speed
Larg. × P × H
5 in. (12.7 cm) × 5 in. (12.7 cm) × 4.4 in. (11.2 cm)
Description générale
Simply insert your slides into the cassettes and close the lid. This Slide Spinner quickly accelerated to 4800 RPM, the optimal speed for quick drying without any splashing. Open the lid and the rotor comes to a stop and the dry slides can be removed. The cassettes catch all of the liquid, and can easily be removed, and the liquid can be poured out.
  • Quickly dries 2 slides
  • Ideal for removing wash buffer from microarrays prior to scanning
Removeable cassettes conveniently catch all liquid
Global Trade Item Number
RéférenceGTIN
Z674672-1EA4061833580721