Principaux documents
Z641812
Schott® tube connectors
bent
Synonyme(s) :
292470504
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
Matériaux
polypropylene
Fabricant/nom de marque
Schott 29 247 05
Disponibilité
available only in EU
Tuyau, D.E. × D.I.
8 mm × 4 mm
Description générale
For screw-cap with aperture GL14. With silicone rubber seal.
Informations légales
Schott is a registered trademark of Schott AG