Principaux documents
Z551732
Sigma-Aldrich
Aldrich® cold-trap bottom
joint: ST/NS 29/32, for dry ice/acetone
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Joint
joint: ST/NS29/32
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC