Principaux documents
Z549185
Sigma-Aldrich
Aldrich® antisplash splash-guard adapter without return hole
female joint: ST/NS 29/32, male joint: ST/NS 24/40, capacity 500 mL
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
Joint
female joint: ST/NS29/32
male joint: ST/NS24/40
Capacité
500 mL
Description générale
Aldrich® antisplash splash-guard adapter without return hole are traps that greatly reduce carry-over from foaming or bumping. Whatever comes up into this trap will stay and not go back to the evaporator flask.
Application
Aldrich® antisplash splash-guard adapter without return hole is ideal for use between the vapor tube and rotary evaporator flask to prevent solvent from being drawn into condenser in case of "bumping".
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC