Principaux documents
Z548839
Sigma-Aldrich
Aldrich® solvent-pouring funnel
stem L × O.D. 100 mm × 28 mm
About This Item
Code UNSPSC :
41121809
Nomenclature NACRES :
NB.43
Tige, L × D.E.
100 mm × 28 mm
Catégories apparentées
Description générale
Unique, closed-end glass funnel has 2 mm diameter holes in the stem permitting the gentle addition of solvents to chromatography columns without disturbing the silica gel/sand layer. The top of the funnel is 75 mm in diameter.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC