Key Documents
Z411833
Acid neutralization traps for Welch® pumps
For Models 1402N and 1376N
About This Item
Code UNSPSC :
41105101
Nomenclature NACRES :
NB.79
Caractéristiques
For Models 1402N and 1376N
Description générale
includes replacement element 1420E-01
Informations légales
Welch is a registered trademark of Gardner Denver Thomas, Inc.