Z341924
IKA® dish attachment for HS/KS 260 basic and control shakers
AS 260.3 dish attachment, 1/cs
Synonyme(s) :
benchtop shakers, lab shakers, laboratory dish shakers, laboratory shakers
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.73
Produits recommandés
Matériaux
foil (slip-resistant)
Conditionnement
case of 1
pack of 1
Fabricant/nom de marque
IKA 3120600
Adéquation
suitable for
Description générale
Note: For smooth movement for cell cultures, nutrient media in Petri dishes, culture bottles and vessels with a low center of gravity; with integrated slip-resistant foil (PP).
Informations légales
IKA is a registered trademark of IKA-Werke GmbH & Co. KG