Principaux documents
Z317586
Duran® vacuum desiccator plate
porcelain plate, I.D. 250 mm
Synonyme(s) :
297256609
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Matériaux
porcelain plate
Fabricant/nom de marque
DURAN®
Schott
Disponibilité
not available in USA and Canada
D.I.
250 mm
Description générale
Perforated plate for vacuum desiccators.
Informations légales
DURAN is a registered trademark of DWK Life Sciences