Principaux documents
Z304026
Quickfit® plain stillhead
Quickfit SH 4/44
Synonyme(s) :
SH4/44
About This Item
Code UNSPSC :
41104812
Nomenclature NACRES :
NB.43
Joint
male joint ST/NS29/32
Fabricant/nom de marque
Quickfit SH 4/44
Disponibilité
not available in USA and Canada, ROW (Available only in EU countries)
not available in USA and Canada
Description générale
With 14/23 thermometer joint.
Informations légales
Quickfit is a registered trademark of SciLabware Limited