Principaux documents
Z303771
Quickfit® three-neck round-bottom flasks with vertical necks
capacity 6,000 mL, center joint: ST/NS 45/40
Synonyme(s) :
FR6L/7S/44P
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass
round bottom flask
three-neck (Vertical)
Joint
center joint: ST/NS45/40
side joint ST/NS29/32
Stérilité
non-sterile
Caractéristiques
closure type Standard Taper Joint
Conditionnement
pack of 1
Fabricant/nom de marque
Quickfit FR6L/7S/44P
Disponibilité
not available in ROW (Available only in EU countries), USA and Canada
not available in USA and Canada
Hauteur
300 mm
Capacité
6,000 mL
Description générale
  • One vertical neck
  • Two parallel necks
Informations légales
Quickfit is a registered trademark of SciLabware Limited