Principaux documents
Z302759
Quickfit® round-bottom flasks
capacity 250 mL, joint: ST/NS 24/29
Synonyme(s) :
FR250/3S
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass
round bottom flask
short
single-neck
Joint
joint: ST/NS24/29
Stérilité
non-sterile
Caractéristiques
closure type Standard Taper Joint
Conditionnement
pack of 1
Fabricant/nom de marque
Quickfit FR250/3S
Disponibilité
not available in USA and Canada
Capacité
250 mL
Description générale
Short neck with ground-glass joint.
Informations légales
Quickfit is a registered trademark of SciLabware Limited