Principaux documents
Z231576
Duran® watch glasses
O.D. 40 mm
Synonyme(s) :
213212408
About This Item
Code UNSPSC :
41121801
Nomenclature NACRES :
NB.43
Fabricant/nom de marque
DURAN®
D.E.
40 mm
Description générale
With fused edges. Manufactured from Duran borosilicate glass type 3.3 for maximum chemical and thermal shock resistance.
Informations légales
DURAN is a registered trademark of DWK Life Sciences