Key Documents
Z155861
Sigma-Aldrich
-NotTranslated-
capacity 500 mL
About This Item
Code UNSPSC :
41121803
Nomenclature NACRES :
NB.43
Matériaux
stainless steel
Caractéristiques
graduations
H × D.E.
117 mm × 95 mm
Capacité
500 mL
Poids
16 oz
Description générale
Standard-weight, spun SS, with handle.