Principaux documents
TEM-HC300CUCC
Sigma-Aldrich
Holey Carbon/Continuous Ultrathin carbon film Supported Copper Grids
size 300 mesh, box of 25 ×
Synonyme(s) :
Holey Carbon Film - Cu, 300 mesh, less than 3 nm, CC
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
MA.24
Conditionnement
box of 25 ×
Taille
300 mesh
Description générale
Holey Carbon Film with a Continuous Layer of Ultrathin Carbon Film (< 3 nm thick) on 300 mesh copper grids.