Principaux documents
TEM-FFT200CU25
Sigma-Aldrich
Formvar Supported Copper Thin Bar Square Mesh
size 200 mesh, box of 25
Synonyme(s) :
FORMVAR SUPPORT FILM THIN BAR SQUARE 200 MESH CU GRIDS
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
MA.24
Conditionnement
box of 25
Taille
200 mesh
Description générale
A 10nm thin film of pure formvar resin supported thin bar square mesh grids. Copper Grid, 200 mesh.