Principaux documents
TEM-FCF100NIUA
Sigma-Aldrich
Ultrathin Formvar/Carbon Supported Nickel Square Mesh
size 100 mesh, box of 50
Synonyme(s) :
FORMVAR/CARBON SQUARE MESH - NI 100 MESH UA 50/BX
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
MA.24
Conditionnement
box of 50
Taille
100 mesh
Catégories apparentées
Description générale
Ultrathin 5-6nm Formvar and 1nm Carbon Supported Square Mesh Grids: Nickel Grid, 100 mesh. A formvar coated grid, stabilized with evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra thin sections.
Global Trade Item Number
RéférenceGTIN
TEM-FCF100NIUA4061842455072