Principaux documents
TEM-CF400-AU-ET
Sigma-Aldrich
Carbon Film Supported Gold Square Mesh
Extra Thick, size 400 mesh, box of 50
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
MA.24
Conditionnement
box of 50
Taille
400 mesh
Description générale
A 20-30 nm thick film of pure carbon supported standard squre mesh grids. Gold grid, 400 mesh.