TEM-CF200AU50
Carbon Film Supported Gold Square Mesh
size 200 mesh, box of 50
Synonyme(s) :
CRBON FILM 200 MESH AU 50/BX
About This Item
Code UNSPSC :
41111761
Conditionnement
box of 50
Taille
200 mesh
Description générale
A 5-6 nm thin film of pure carbon supported standard squre mesh grids. Gold grid, 200 mesh.