Principaux documents
TEM-CF150-AU-TH
Sigma-Aldrich
Carbon Film Supported Gold Square Mesh
Thick, size 150 mesh, box of 50
About This Item
Code UNSPSC :
41111761
Conditionnement
box of 50
Taille
150 mesh
Description générale
A 10 nm thick film of pure carbon supported standard squre mesh grids. Gold grid, 150 mesh.