Principaux documents
SYNF419900
Synthware two neck round bottom flask with angled side neck
100 mL, center joint: ST/NS 19/22, side joint: ST/NS 19/22
Synonyme(s) :
Synthware F419900, glass flask, 2-neck flask
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.51
Matériaux
borosilicate glass flask
round bottom
two-neck (angled side neck)
Joint
center joint: ST/NS19/22
side joint: ST/NS19/22
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F419900
Capacité
100 mL
100 mL
Description générale
Round bottom with joints. Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
Informations légales
Synthware is a trademark of Kemtech America Inc.