Principaux documents
SLW1517/12
Pyrex® Media-Lab Bottles, with-out cap and pouring ring, with printed trace code
capacity 2000 mL
Synonyme(s) :
Pyrex, SciLabware 1517/12
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.17
Capacité:
2000 mL
Diam. × H:
136 mm × 260 mm
Matériaux:
borosilicate glass bottle
round bottle
white graduation
Matériaux
borosilicate glass bottle
round bottle
white graduation
Joint
Joints threaded neck (GL45)
Caractéristiques
graduations
without cap
with printed trace code
without pouring ring
Conditionnement
pack of 10 ea
Fabricant/nom de marque
SciLabware 1517/12
Capacité
2000 mL
Diam. × H
136 mm × 260 mm
Description générale
  • Bottles only. Supplied without cap or pouring ring
  • Complies with ISO 4796
  • Printed with Trace Code for downloadable batch certificate
  • Batch certificate available to download from www.scilabware.com
  • Sloping shoulders for smooth pouring action
  • White graduations and marking spot
  • Manufactured from temperature and chemically resistant borosilicate glass
Caractéristiques et avantages
• Bottles only. Supplied without cap or pouring ring.• Complies with ISO 4796.• Printed with Trace Code for downloadable batch certificate.• Sloping shoulders for smooth pouring action.• White graduations and marking spot.• Manufactured from temperature and chemically resistant borosilicate glass.• Choice of caps and pouring rings available separately.• The 25 ml size bottle features a glass lip and does not require a pouring ring.
Informations légales
PYREX is a registered trademark of Corning, Inc.