Principaux documents
NORTCM300
Bruker tapered PTFE caps
for NORS305001
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
SB.32
Caractéristiques
for NORS305001
Conditionnement
pack of 5 ea
Fabricant/nom de marque
Norell TCM300
Description générale
Bruker Match tube caps are specially machined from PTFE fluoropolymer, are color coded by tube size, and correspond to the color of the tube clamp in the Match Insert Assembly, This enables quick identification of matching components. The tube cap incorporates a slightly tapered inner diameter. Initially, the cap aligns and pushes easily onto the tube, it then becomes progressively tighter as the tube reaches full depth within the cap, creating a positive seal to preserve volatile sample solutions throughout short or long term storage.
Informations légales
Match is a trademark of Bruker Biospin Corp.