Principaux documents
DWKW242824-A
Wheaton® Dropping Bottle
natural bottle, capacity 10 mL, case of 1000
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.17
Matériaux
natural bottle
Conditionnement
pkg of 1 packs
case of 1000
Fabricant/nom de marque
WHEATON® W242824-A
Capacité
10 mL
Application
Opthalmic, IVD Kits, Other
Caractéristiques et avantages
  • Bottle made of natural or white LDPE (Low Density Polyethylene
  • White LDPE bottle helps protect UV light sensitive samples
  • Dropper tip caps and tips sold separately
  • Packaged in cases of 100 or 1000
Informations légales
WHEATON is a registered trademark of Wheaton Industries, Inc.