Principaux documents
DWK218606904
DURAN® wide mouth laboratory bottle
without cap, round glass bottle, bottle capacity (3,500 mL), non-sterile
Synonyme(s) :
218606904
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.17
Matériaux
round glass bottle
Stérilité
non-sterile
Caractéristiques
without cap
Conditionnement
case of 1 ea
Fabricant/nom de marque
DURAN® 218606904
Contenance du flacon
(3,500 mL)
Raccords
GL 80 cap
GL 80 neck
Description générale
Easy access to the bottle contents due to the wide diameter of the bottle neck. Optimum filling and pouring and particularly easy cleaning. Spoons, spatulas and tweezers can be manipulated with ease. Even large-volume funnels can be inserted into the bottles without any difficulty. Stability due to the large base area. Easy-to-read scale with graduation marks to estimate the contentFired-on, highly durable white ceramic. Complete with blue screw cap (PP, integral lip seal) and pouring ring (PP) for drip-free pouring and clean, safe working
Informations légales
DURAN is a registered trademark of DWK Life Sciences