Principaux documents
DWK212173606
DURAN® Super duty Erlenmeyer narrow-neck flasks
flask capacity (250 mL)
Synonyme(s) :
212173606
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
flat
Mode de stérilisation compatible
autoclavable compatible
Caractéristiques
without cap
Conditionnement
case of 10 ea
Fabricant/nom de marque
DURAN® 212173606
Contenance du flacon/de la fiole
(250 mL)
Description générale
Higher impact strength, mechanical strength and stability achieved by reinforced rim. Economical due to longer life span and outstanding corrosion protection. White easy-to-read scale with graduation marks to estimate the content. Fired-on, highly durable white ceramic print.
Autres remarques
Informations légales
DURAN is a registered trademark of DWK Life Sciences