Principaux documents
CLS5100500
Pyrex® wide-mouth Erlenmeyer flask
capacity 500 mL
Synonyme(s) :
conical flasks, erlenmeyer flasks, flask erlenmeyer, flasks erlenmeyer, Corning® 5100
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass
wide-mouth
Joint
joint stopper10
Stérilité
non-sterile
Caractéristiques
cap: no
subdivision 50
Conditionnement
case of 36
pack of 6
Fabricant/nom de marque
Corning 5100-500
Plage de mesure du volume
100-500 mL
Capacité
500 mL
Description générale
  • Pyrex® wide-mouth Erlenmeyer flasks are made of borosilicate glass for optical clarity and mechanical strength.
  • Flasks have durable white enamel graduations to show approximate capacity and an extra-large marking spot.
  • Features a heavy-duty rim.
  • Ideal for use as a titration flask.
Heavy-duty rim. Especially recommended as a titration flask.
Autres remarques
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.