Principaux documents
CAD7915
Animal feeding needle, non-sterile, reusable, type 304 SS
curved, size 16G, L × diam. 2 in. × 3 mm , ball
About This Item
Code UNSPSC :
42142609
Nomenclature NACRES :
NB.03
Taille
16G
Conditionnement
pkg of 12 ea
L × diam.
2 in. × 3 mm , ball
Caractéristiques et avantages
  • Fixed shape from stainless steel tubing
  • Stainless steel balls
  • Can be cleaned and reused as needed
  • Non-sterile