Key Documents
BR705430
BRAND® Transferpette® multichannel, electronic pipette
AC/DC input 230 V AC, Australian plug, 8-channel, volume 1-20 μL
Synonyme(s) :
Air displacement pipettes, Air interface pipettes, Microliter pipettes, Multichannel pipettes, Pipettes, Piston-operated pipettes, Transferpette® electronic
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.01
Caractéristiques
8-channel
electronic
Conditionnement
pack of 1 ea
Fabricant/nom de marque
BRAND 705430
Volume
1-20 μL
Alimentation par piles
rechargeable
Description générale
Transferpette electronic micropipettes combine the globally proven properties of mechanical piston operated air displacement micropipettes by BRAND with the advantages of an electronic device. The Transferpette electronic is the first micropipette to be awarded the "Ergonomics Approved" certificate from the TÜV Rheinland. The special FKM V-rings allow effortless mounting and easy ejection of virtually all commercially available pipette tips.
  • Motor-driven to reduce the risk of RSI and imprecision/inaccuracy caused by operator variability
  • Five modes: Pipetting, Reverse pipetting, Mixing, Dispensing, Gel-electrophoresis
  • Easy and rapid calibration without tools
  • The pipette shaft can be unscrewed and is entirely autoclavable (121 °C).
For pipettes up to 300 µl, includes: Transferpette®-8/-12 electronic, performance certificate, battery, power adapter, device stand, TipBox, refill pack, reagent reservoir, installation tool, silicone grease, and 1 set of sealing rings
For pipettes up to 1250 µl, includes: Transferpette®-8/-12 electronic, performance certificate, battery, power adapter, support frame, shelf mount, 2x TipBox, reagent reservoir, installation tool, grease and 1 set of sealing rings.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG
Transferpette is a registered trademark of BRAND GMBH + CO KG
Certificats d'analyse (COA)
Lot/Batch Number
Désolés, nous n'avons pas de COA pour ce produit disponible en ligne pour le moment.
Protocoles
This article and video demonstrate how to change the V-Rings and O-Rings on the tip cones of any Transferpette® Multichannel Pipette.
This article describes easy calibration techniques for mechanical and electronic liquid handling instruments from BRAND®.
This article describes easy calibration techniques for mechanical and electronic liquid handling instruments from BRAND®.