Principaux documents
BR150045
BRAND® watch glass
soda-lime glass, diam. 90 mm , ground edges stress-reduced
Synonyme(s) :
Dishes, Watch glasses
About This Item
Code UNSPSC :
41121801
Nomenclature NACRES :
NB.43
Matériaux
soda-lime glass
Fabricant/nom de marque
BRAND 150045
Diamètre
90 mm , ground edges stress-reduced
Description générale
The BRAND® watch glass is made from soda-lime glass and complies with DIN 12341 standards. It features ground edges for safe handling and a refined finish. This watch glass is stress-reduced to minimize the risk of shattering, enhancing safety during laboratory use. Ideal for covering beakers or evaporating dishes, it provides an effective solution for preventing contamination and evaporation of samples. Its durable design makes it a reliable tool in various laboratory applications, ensuring optimal performance in your experiments.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG