Principaux documents
BMSC2417EUK
Benchmark MC-24 Touch High Speed Microcentrifuge with COMBI-Rotor
AC/DC input 230 V, UK plug
Synonyme(s) :
High speed centrifuge, laboratory centrifuge, benchtop centrifuge, bendh top centrifuge, lab centrifuge
About This Item
Code UNSPSC :
40161701
Nomenclature NACRES :
NB.13
Caractéristiques
refrigerated: no
to hold 24 x 1.5/2.0ml tubes and 2 strips of 0.2ml tubes (16 x 0.2ml)
Fabricant/nom de marque
Benchmark Scientific C2417-E-UK
Paramètres
13,500 rpm speed
Description générale
Benchmark′s MC-24 Touch is the first microcentrifuge in its class to feature touch screen control. In addition to the next generation control panel, the centrifuge also offers high speed (up to 13,500 rpm/16,800xg) and a unique COMBI-Rotor that accepts both standard microtubes (1.5/2.0ml)and PCR tubes/strips (0.2ml), making this the perfect partner for any molecular biology laboratory.
The large, color touch screen shows actual speed (in rpm or rcf) along with set and remaining time. Up to 9 speed/time profiles can be stored in memory and the home screen displays a "mySpin" icon for instant recalls a designated favorite profile.
Supplied with Benchmark’s unique , the MC-24Touch will accept 24 x 1.5/2.0ml tubes and 2 strips of 0.2ml tubes (16 x 0.2ml)without requiring separate adapters. Samples are kept cool, even during extended runs at the maximum speed setting by a proprietary air flow system that moves air evenly and quietly throughout the chamber.
The MC-24 Touch is cold room safe
Informations légales
COMBI-Rotor is a trademark of Benchmark Scientific, Inc.