MilliporeSigma
Toutes les photos(5)
Principaux documents
BAF369740420
SP Bel-Art® Sterileware Powder Lance Samplers
white high-density polyethylene, capacity 75 mL (2.5 oz), L 15.75 in. (40 cm), sterile, pack of 20 ea (individually wrapped)
Synonyme(s) :
sampler
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.35
Matériaux
white high-density polyethylene
Stérilité
sterile
Conditionnement
pack of 20 ea (individually wrapped)
Fabricant/nom de marque
Bel-Art F36974-0420
Longueur
15.75 in. (40 cm)
Capacité
75 mL (2.5 oz)
Catégories apparentées
Description générale
A simple insert, rotate, and withdraw motion allows cross sectional sampling of non-flowing, bulk powders. Available in three lengths.
Caractéristiques et avantages
  • Open blade works well with sticky, cohesive powders
  • Rounded edges help hold sample in place
  • Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
  • High density polyethylene (HDPE) lance is heat sealed in individual polyethylene bag
  • Gamma irradiated sterile (SAL 10^-6)
  • Ready for one time use and disposal
  • Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization
Informations légales
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Sterileware is a trademark of SP Scienceware, a division of SP