Principaux documents
935077
Sigma-Aldrich
VC-2 Voltammetry cell
5-15 mL
Synonyme(s) :
MF-1052
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
NA.21
Plage de mesure
5-15 mL
Description générale
VC-2 Voltammetry Cell (5-15 mL of solution) includes PTFE cap, Platinum auxiliary electrode - 99.95% pure, 0.5 mm diameter,Glass vial - shell type, and tubing 0.025" ID x 1/6" OD (1 ft).This Electrochemical cellware is ideal for testing 5-15 mL of solution