MilliporeSigma
TLC Analysis of Caffeine in Coffee Samples, Quantification of Analyte Concentration
TLC Analysis of Caffeine in Coffee Samples, Quantification of Analyte Concentration application for TLC
Materials
accessory
Product No.
Description
Pricing
Bottle-type sprayer
capacity 240 mL
Rectangular TLC developing tanks, complete
L × H × W 27.0 cm × 26.5 cm × 7.0 cm
Sigma TLC spray box
Spectroline® E-Series UV lamp
output8 W, AC/DC input 115 V AC
Rectangular TLC developing tanks, complete
L × H × W 7.5 cm × 15.5 cm × 8.0 cm
Hamilton® TLC syringes
1702, volume 25 μL, needle size 22s ga, needle L 51 mm (2 in.),
coated tip
Spectroline® E-Series UV lamp
output6 W, AC/DC input 120 V AC
Hamilton® TLC syringes
1705, volume 50 μL, needle size 22 ga, needle L 51 mm (2 in.),
coated tip
Hamilton® TLC syringes
1701, volume 10 μL, needle size 26s ga, needle type, fixed, needle L 51 mm (2 in.),
coated tip
Rectangular TLC developing tanks, complete
L × H × W 17.5 cm × 16.0 cm × 8.2 cm
Spectroline® E-Series UV lamp
output6 W, AC/DC input 230 V AC
Spectroline® E-Series UV lamp
output4 W, AC/DC input 230 V AC
Spectroline® E-Series UV lamp
output8 W, AC/DC input 230 V AC
Latch-lid TLC developing chambers
For use with 10 cm × 10 cm plates
Hamilton® TLC syringes
1710, volume 100 μL, needle size 22 ga, needle L 51 mm (2 in.),
coated tip
Latch-lid TLC developing chambers
For use with 20 cm × 20 cm plates
Spectroline® E-Series UV lamp
output4 W, AC/DC input 115 V AC
Kimble® microcapillary pipettes
Kimble® microcapillary pipettes
Kimble® microcapillary pipettes
capacity 20 μL, accuracy: ±0.5%
Reagent sprayer for TLC plates
volume 250 mL, Bottle-Type
CONDITIONS
sample preparation
The boiled coffee samples were extracted and filtered with a 0.45 μm Millex syringe filter.
plate
HPTLC Silica gel 60 F254, 20 cm x 10 cm (1.05642.0001)
eluent
[A] acetonitrile; [B] methanol; [C] n-Heptane; (6:4:1, A:B:C)
migration distance
5 cm
sample
0.1 μL to 2.0 μL
detector
UV, 254 nm
Description
Analysis Note
Abstract: HPTLC (High Performance Thin Layer Chromatography) is a very useful analytical technique with very low sample preparation. Shown in the example: the quantification of caffeine in different coffee samples. The boiled coffee samples
were extracted and filtered with a 0.45 μm syringe filter. The plate was pre-conditioned with the mobile phase. After chromatographic separation the plate was scanned with a TLC-Scanner and subsequently quantified.
Results: Caffeine was detected under UV light (254 nm, hRf = 56, Fig. 1), (quantification information Fig. 2). A triple determination of each sample is possible in parallel.
Conclusion: These results demonstrate that caffeine can be easily quantified by TLC with a three-fold determination and minimal sample preparation.