MilliporeSigma
LC/MS Analysis of Sertraline and Norsertraline on Ascentis® Express RP-Amide
LC/MS Analysis of Sertraline and Norsertraline on Ascentis® Express RP-Amide application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express RP-Amide, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 10 cm × 3 mm
CONDITIONS
column
Ascentis Express RP Amide, 10 cm x 3.0 mm I.D., 2.7 μm particles (53918-U)
mobile phase
[A] 10 mM ammonium formate, water, pH 3.6; [B] 10 mM ammonium formate, acetonitrile, pH 3.6; (65:35, A:B)
flow rate
0.6 mL/min
column temp.
55 °C
detector
ESI(+), 100-1000 m/z
injection
2 μL
sample
200 ng/mL in 25:75, water: acetonitrile
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany