MilliporeSigma
HPLC Analysis of Peroxidase on TSKgel® G3000SWXL
HPLC Analysis of Peroxidase on TSKgel® G3000SWXL application for HPLC
Materials
Application
Product No.
Description
Pricing
TSKgel® G3000SWXL HPLC Column
phase diol, L × I.D. 30 cm × 7.8 mm, 5 μm particle size
CONDITIONS
column
TSKgel G3000SWXL, 30 cm × 7.8 mm I.D., 5 μm particles (808541)
mobile phase
0.3M sodium chloride/0.05M phosphate buffer, pH 7.0
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 220 nm
injection
100 μL crude peroxidase preparation from Japanese horseradish (0.15mg)
Description
Legal Information
TSKgel is a registered trademark of Tosoh Corporation