MilliporeSigma
HPLC Analysis of Methadone Enantiomers and Methadone Metabolite EDDP on Astec® CYCLOBOND I 2000 HP-RSP
HPLC Analysis of Methadone Enantiomers and Methadone Metabolite EDDP on Astec® CYCLOBOND I 2000 HP-RSP application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Astec® CYCLOBOND I 2000 HP-RSP Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
CONDITIONS
column
CYCLOBOND I 2000 HP-RSP, 25 cm x 4.6 mm I.D., 5 μm particles (24024AST)
mobile phase
[A] 0.1% acetic acid; [B] acetonitrile; (80:20, A:B)
flow rate
0.8 mL/min
detector
UV, 292 nm
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
null