MilliporeSigma
HPLC Analysis of Lysozyme on SRT® SEC-300, 30 cm x 7.8 mm I.D., 5 μm particles
HPLC Analysis of Lysozyme on SRT® SEC-300, 30 cm x 7.8 mm I.D., 5 μm particles application for HPLC
Materials
standard
Product No.
Description
Pricing
Lysozyme from chicken egg white
powder or granules, ≥90 %, ≥39,000 units/mg protein
CONDITIONS
column
SRT SEC-300, 30 cm x 7.8 mm I.D., 5 μm particles (Z777051)
mobile phase
50 mM phosphate buffered saline, 200 mM Na2SO4, pH 7.0
flow rate
1 mL/min
column temp.
ambient
detector
UV, 214 nm
injection
5 μL
sample
1 mg/mL in 50 mM phosphate buffered saline
Description
Analysis Note
The positively-charged protein, lyzoyme (pI=11), exhibits excellent peak shape and recovery on SRT SEC-300 columns due to its highly-inert surface. Low protein recovery is usually attributed to 1) hydrophobic interactions with the stationary phase and 2) electrostatic interactions with the silica suface. SRT SEC-300 surface is hydrophilic and densely bonded to silica, therefore providing high mass recovery and good peak shape.
Legal Information
SRT is a registered trademark of Sepax Technologies