MilliporeSigma
HPLC Analysis of Tramadol Enantiomers on Astec® CYCLOBOND I 2000 DMP
HPLC Analysis of Tramadol Enantiomers on Astec® CYCLOBOND I 2000 DMP application for HPLC
Materials
standard
Product No.
Description
Pricing
Tramadol hydrochloride
≥99.0% (HPLC)
CONDITIONS
column
CYCLOBOND I 2000 DMP, 25 cm x 4.6 mm I.D., 5 μm particles (20724AST)
mobile phase
[A] 0.1% triethylamine acetate, pH 5.0; [B] acetonitrile; (80:20, A:B)
flow rate
1.2 mL/min
column temp.
23 °C
detector
UV, 254 nm
injection
5 μL
sample
5 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
null