MilliporeSigma
HPLC Analysis of Bendroflumethiazide Enantiomers on Astec® CHIROBIOTIC® T
HPLC Analysis of Bendroflumethiazide Enantiomers on Astec® CHIROBIOTIC® T application for HPLC
Materials
standard
Product No.
Description
Pricing
Bendroflumethiazide
analytical standard
CONDITIONS
column
CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 μm particles (12024AST)
mobile phase
[A] water; [B] ethanol; (70:30, A:B)
flow rate
1 mL/min
column temp.
23 °C
detector
UV, 215 nm
injection
5 μg/mL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC