MilliporeSigma
HPLC Analysis of Withania (Aswagandha) Standard on Ascentis® Express F5
HPLC Analysis of Withania (Aswagandha) Standard on Ascentis® Express F5 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express F5, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 10 cm × 2.1 mm
CONDITIONS
column
Ascentis Express F5, 10 cm x 2.1 mm I.D., 2.7 μm particles (53569-U)
mobile phase
[A] water; [B] acetonitrile
gradient
20 to 100% B in 10 min: held at 100% B for 0.5 min
flow rate
0.3 mL/min
pressure
2756 psi (190 bar)
column temp.
35 °C
detector
UV, 227 nm
injection
5 μL
sample
20 μg/mL in 80:20, water:methanol
Description
Analysis Note
This application demonstrates the suitability of Ascentis Express F5 for the efficient separation of withania standards.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany