HPLC Analysis of Selegiline and Amphetamine Metabolites on Discovery® HS F5
![HPLC Analysis of Selegiline and Amphetamine Metabolites on Discovery® HS F5 application for HPLC](/deepweb/assets/sigmaaldrich/product/images/216/298/4d410cc2-693a-4c20-b288-557b0d7d3519/800/4d410cc2-693a-4c20-b288-557b0d7d3519.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Discovery HS F5, 5 cm × 4.6 mm I.D., 5 μm particles (567513-U)
mobile phase
[A] water (10 mM ammonium acetate, pH 4.0 with acetic acid: [B] acetonitrile (v/v), (25:75, A:B)
flow rate
1.0 mL/min
column temp.
40 °C
detector
UV, 210 nm
injection
10 μL
sample
50 μg/mL each in mobile phase
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany