MilliporeSigma
HPLC Analysis of Organic Acids on Ascentis® Express RP-Amide
HPLC Analysis of Organic Acids on Ascentis® Express RP-Amide application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express RP-Amide, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 10 cm × 2.1 mm
standard
Product No.
Description
Pricing
Citric acid
99%
Fumaric acid
≥99.0% (T)
L-(+)-Tartaric acid
≥99.5%
Acrylic acid
anhydrous, contains 200 ppm MEHQ as inhibitor, 99%
Lactic acid
Pharmaceutical Secondary Standard; Certified Reference Material
CONDITIONS
column
Ascentis Express RP-Amide, 10 cm x 2.1 mm I.D., 2.7 μm particles (53913-U)
mobile phase
[A] 0.1% (v/v) TFA in [B] water
flow rate
0.3 mL/min
column temp.
35 °C
detector
UV, 210 nm
injection
1 μL
sample
0.2 - 4 g/L in mobile phase
Description
Analysis Note
This application demonstrates the suitability of Ascentis Express RP-Amide for the analysis of organic acids under 100% aqueous conditions.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany