MilliporeSigma
HPLC Analysis of FMOC-Threonine Enantiomers on Astec® CHIROBIOTIC® T
HPLC Analysis of FMOC-Threonine Enantiomers on Astec® CHIROBIOTIC® T application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIROBIOTIC® T Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
CONDITIONS
column
CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 μm particles (12024AST)
mobile phase
(A) 20 mM ammonium acetate, pH 4.1; (B) methanol; (60:40, A:B)
flow rate
0.8 mL/min
column temp.
25 °C
detector
UV, 260 nm
injection
1 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC