MilliporeSigma
HPLC Analysis of 3a,4,5,6-Tetrahydrosuccinimido[3,4-b]acenaphthen-10-one Enantiomers on Astec® CHIROBIOTIC® V
HPLC Analysis of 3a,4,5,6-Tetrahydrosuccinimido[3,4-b]acenaphthen-10-one Enantiomers on Astec® CHIROBIOTIC® V application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIROBIOTIC® V Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
CONDITIONS
column
CHIROBIOTIC V, 25 cm x 4.6 mm I.D., 5 μm particles (11024AST)
mobile phase
[A] hexane; [B] ethanol; (75:25, A:B)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 254 nm
injection
1 μL
sample
4 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC