MilliporeSigma
US EPA Method TO-13: GC Analysis of PAHs on SLB®-5ms
US EPA Method TO-13: GC Analysis of PAHs on SLB®-5ms suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SLB®-5ms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
SLB®-5ms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.50 μm
SLB®-5ms Capillary GC Column
L × I.D. 20 m × 0.18 mm, df 0.18 μm
accessory
Product No.
Description
Pricing
Supeltex M-2B Double-Tapered Ferrule
I.D. 0.4 mm, 0.10-0.25 mm to like I.D. Column, pkg of 2 ea
Capillary Column Butt Connector
body only (ferrules not included)
guard column
Product No.
Description
Pricing
Intermediate Polar Fused Silica Tubing
L × I.D. 5.0 m × 0.25 mm
related product
Product No.
Description
Pricing
ORBO 2000 Precleaned Large PUF Cartridge
O.D. × L 6 cm × 7.6 cm, pkg of 1 ea
Glass Cartridge and Screen for Large PUF Plug
pkg of 1 ea
Stainless Steel Screen
ORBO 2000 Precleaned Large PUF Plug, Replacement
O.D. × L 6 cm × 7.6 cm, pkg of 1 ea
Supelpak-2
matrix styrene-divinylbenzene, 20-60 mesh, jar of 100 g
Inlet Liner, Split/Splitless Type, Straight Fast FocusLiner Design (wool packed)
pkg of 1 ea
standard
Product No.
Description
Pricing
Coronene
purified by sublimation, 99%
Benzo[e]pyrene
analytical standard
Perfluorotributylamine (PFTBA)
analytical standard
EPA 8270 GC-MS Tuning Solution II
certified reference material, 1000 μg/mL each component in dichloromethane
Perylene solution
certified reference material, 2000 μg/mL in dichloromethane
ORBO 2500 Precleaned Large PUF/Amberlite XAD®-2/PUF Cartridge
O.D. × L 6.5 cm × 125 mm, pkg of 1 ea
EPA 8270 Semivolatile Internal Standard Mix
certified reference material, TraceCERT®, 2000 μg/mL each component in methylene chloride, ampule of 2 × 1 mL
Polynuclear Aromatic Hydrocarbons Mix
certified reference material, TraceCERT®, 2000 μg/mL each component in methylene chloride: benzene (1:1)
EPA 610 Polynuclear Aromatic Hydrocarbons Mix
certified reference material, in methanol: methylene chloride (1:1) (varied concentration)
CONDITIONS
column
SLB-5ms, 30 m x 0.25 mm I.D., 0.25 μm (28471-U)
oven
60 °C (1 min), 10 °C/min to 340 °C (5 min)
inj. temp.
250 °C
MSD interface
330 °C 300 °C / 200 °C (source/quads)
scan range
m/z 40-450
carrier gas
helium, 1.5 mL/min. constant
injection
1.5 μL, pulsed (25 psi until 0.2 min) splitless (0.75 min)
liner
2 mm I.D. FocusLiner liner (2879601-U)
sample
22 component TO-13 standard plus 5 internal standards at 500 ppb in hexane:ethyl ether (90:10)
Description
Legal Information
FocusLiner is a trademark of SGE Analytical Science Pty Ltd.
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany